accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNOOH

CAS RN 898289-01-5

IUPAC Name: 3-(piperidin-1-yl)pyridine-2-carboxylic acid
Synonyms: 3-(piperidin-1-yl)pyridine-2-carboxylic acid
Molecular Weight (g/mol): 206.25
Molecular Formula: C11H14N2O2
InChi Key: QIWMAMIMWHCYGZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=CC=N1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results