accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 898289-24-2

SNNO

CAS RN 898289-24-2

IUPAC Name: 2-(thiomorpholin-4-yl)pyridine-4-carbaldehyde
Synonyms: 2-(thiomorpholin-4-yl)pyridine-4-carbaldehyde
Molecular Weight (g/mol): 208.28
Molecular Formula: C10H12N2OS
InChi Key: XESFZSHCANCGBY-UHFFFAOYSA-N
SMILES: O=CC1=CC(=NC=C1)N1CCSCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results