accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 898289-25-3

SNNOH

CAS RN 898289-25-3

IUPAC Name: [2-(thiomorpholin-4-yl)pyridin-4-yl]methanol
Synonyms: [2-(thiomorpholin-4-yl)pyridin-4-yl]methanol
Molecular Weight (g/mol): 210.3
Molecular Formula: C10H14N2OS
InChi Key: KDJBEJVGUFHDPW-UHFFFAOYSA-N
SMILES: OCC1=CC(=NC=C1)N1CCSCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results