accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOCl

CAS RN 89901-02-0

IUPAC Name: methyl 4'-chloro-[1,1'-biphenyl]-4-carboxylate
Synonyms: methyl 4'-chloro-[1,1'-biphenyl]-4-carboxylate
Molecular Weight (g/mol): 246.69
Molecular Formula: C14H11ClO2
InChi Key: UJIUMBFXWGXMOV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results