accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 90-04-0

IUPAC Name: 2-methoxyaniline
Synonyms: O-anisidine o-anisidine antimonyl tartrate
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: VMPITZXILSNTON-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1N
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results