accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHO

CAS RN 90-05-1

IUPAC Name: 2-methoxyphenol
Synonyms: guaiacol catechol, methyl guaiastil creodon guaiacol (natural) guaicol guaiacol (substance) guasol guajol o-methyl catechol
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: LHGVFZTZFXWLCP-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (7)
  • (7)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (7)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)