accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 90-13-1

IUPAC Name: 1-chloronaphthalene
Synonyms: 1-chloronaphthalene
Molecular Weight (g/mol): 162.62
Molecular Formula: C10H7Cl
InChi Key: JTPNRXUCIXHOKM-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (3)
Grade 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results