accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 90-15-3

IUPAC Name: naphthalen-1-ol
Synonyms: naphthol ion(1+) 1-naphthol basf ursol ern fourrine ern 1-naphthol, 2-(13)C-labeled nako trb CI oxidation base 33 zoba ern durafur developer D 1-(14)c-labeled 1-naphthol
Molecular Weight (g/mol): 144.17
Molecular Formula: C10H8O
InChi Key: KJCVRFUGPWSIIH-UHFFFAOYSA-N
SMILES: OC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (8)
  • (6)
Quantity 
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (8)
  • (6)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)