accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NCH3ONCH3H3C

CAS RN 90-93-7

IUPAC Name: 4-[4-(diethylamino)benzoyl]-N,N-diethylaniline
Synonyms: 4-[4-(diethylamino)benzoyl]-N,N-diethylaniline
Molecular Weight (g/mol): 324.47
Molecular Formula: C21H28N2O
InChi Key: VYHBFRJRBHMIQZ-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(CC)CC
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results