accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrON

CAS RN 90004-90-3

IUPAC Name: 2-(2-bromophenoxy)acetonitrile
Synonyms: 2-(2-bromophenoxy)acetonitrile
Molecular Weight (g/mol): 212.05
Molecular Formula: C8H6BrNO
InChi Key: VNMBGABMQPZLCO-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1OCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results