CAS RN 9003-39-8
Polyvinylpyrrolidone, M.W. 40,000
CAS: 9003-39-8 Molecular Formula: (C6H9NO)n Molecular Weight (g/mol): 111.14 MDL Number: MFCD01076626 InChI Key: WHNWPMSKXPGLAX-UHFFFAOYSA-N Synonym: n-vinyl-2-pyrrolidone,n-vinylpyrrolidone,1-vinyl-2-pyrrolidone,1-vinylpyrrolidin-2-one,n-vinyl-2-pyrrolidinone,vinylpyrrolidone,2-pyrrolidinone, 1-ethenyl,1-vinyl-2-pyrrolidinone,n-vinylpyrrolidinone,1-vinylpyrrolidone PubChem CID: 6917 ChEBI: CHEBI:82551 IUPAC Name: 1-ethenylpyrrolidin-2-one SMILES: *-CC(-*)N1CCCC1=O
Polyvinylpyrrolidone, average M.W. 1.300.000, K85-95
CAS: 9003-39-8 Molecular Formula: (C6H9NO)n Molecular Weight (g/mol): 111.14 MDL Number: MFCD01076626 InChI Key: WHNWPMSKXPGLAX-UHFFFAOYSA-N Synonym: n-vinyl-2-pyrrolidone,n-vinylpyrrolidone,1-vinyl-2-pyrrolidone,1-vinylpyrrolidin-2-one,n-vinyl-2-pyrrolidinone,vinylpyrrolidone,2-pyrrolidinone, 1-ethenyl,1-vinyl-2-pyrrolidinone,n-vinylpyrrolidinone,1-vinylpyrrolidone PubChem CID: 6917 ChEBI: CHEBI:82551 IUPAC Name: 1-ethenylpyrrolidin-2-one SMILES: *-CC(-*)N1CCCC1=O
Polyvinylpyrrolidone, average M.W. 8.000, K16-18
CAS: 9003-39-8 Molecular Formula: (C6H9NO)n Molecular Weight (g/mol): 111.14 MDL Number: MFCD01076626 InChI Key: WHNWPMSKXPGLAX-UHFFFAOYSA-N Synonym: n-vinyl-2-pyrrolidone,n-vinylpyrrolidone,1-vinyl-2-pyrrolidone,1-vinylpyrrolidin-2-one,n-vinyl-2-pyrrolidinone,vinylpyrrolidone,2-pyrrolidinone, 1-ethenyl,1-vinyl-2-pyrrolidinone,n-vinylpyrrolidinone,1-vinylpyrrolidone PubChem CID: 6917 ChEBI: CHEBI:82551 IUPAC Name: 1-ethenylpyrrolidin-2-one SMILES: *-CC(-*)N1CCCC1=O
Polyvinylpyrrolidone, M.W. 10,000
CAS: 9003-39-8 Molecular Formula: (C6H9NO)n Molecular Weight (g/mol): 111.14 MDL Number: MFCD01076626 InChI Key: WHNWPMSKXPGLAX-UHFFFAOYSA-N Synonym: n-vinyl-2-pyrrolidone,n-vinylpyrrolidone,1-vinyl-2-pyrrolidone,1-vinylpyrrolidin-2-one,n-vinyl-2-pyrrolidinone,vinylpyrrolidone,2-pyrrolidinone, 1-ethenyl,1-vinyl-2-pyrrolidinone,n-vinylpyrrolidinone,1-vinylpyrrolidone PubChem CID: 6917 ChEBI: CHEBI:82551 IUPAC Name: 1-ethenylpyrrolidin-2-one SMILES: *-CC(-*)N1CCCC1=O
Polyvinylpyrrolidone, average M.W. 58,000
CAS: 9003-39-8 Molecular Formula: (C6H9NO)n Molecular Weight (g/mol): 111.14 MDL Number: MFCD01076626 InChI Key: WHNWPMSKXPGLAX-UHFFFAOYSA-N Synonym: n-vinyl-2-pyrrolidone,n-vinylpyrrolidone,1-vinyl-2-pyrrolidone,1-vinylpyrrolidin-2-one,n-vinyl-2-pyrrolidinone,vinylpyrrolidone,2-pyrrolidinone, 1-ethenyl,1-vinyl-2-pyrrolidinone,n-vinylpyrrolidinone,1-vinylpyrrolidone PubChem CID: 6917 ChEBI: CHEBI:82551 IUPAC Name: 1-ethenylpyrrolidin-2-one SMILES: *-CC(-*)N1CCCC1=O
Polyvinylpyrrolidone, average M.W. 50.000, K30
CAS: 9003-39-8 Molecular Formula: (C6H9NO)n Molecular Weight (g/mol): 111.14 MDL Number: MFCD01076626 InChI Key: WHNWPMSKXPGLAX-UHFFFAOYSA-N Synonym: n-vinyl-2-pyrrolidone,n-vinylpyrrolidone,1-vinyl-2-pyrrolidone,1-vinylpyrrolidin-2-one,n-vinyl-2-pyrrolidinone,vinylpyrrolidone,2-pyrrolidinone, 1-ethenyl,1-vinyl-2-pyrrolidinone,n-vinylpyrrolidinone,1-vinylpyrrolidone PubChem CID: 6917 ChEBI: CHEBI:82551 IUPAC Name: 1-ethenylpyrrolidin-2-one SMILES: *-CC(-*)N1CCCC1=O
Polyvinylpyrrolidone, M.W. 1,300,000
CAS: 9003-39-8 Molecular Formula: (C6H9NO)n Molecular Weight (g/mol): 111.14 MDL Number: MFCD01076626 InChI Key: WHNWPMSKXPGLAX-UHFFFAOYSA-N Synonym: n-vinyl-2-pyrrolidone,n-vinylpyrrolidone,1-vinyl-2-pyrrolidone,1-vinylpyrrolidin-2-one,n-vinyl-2-pyrrolidinone,vinylpyrrolidone,2-pyrrolidinone, 1-ethenyl,1-vinyl-2-pyrrolidinone,n-vinylpyrrolidinone,1-vinylpyrrolidone PubChem CID: 6917 ChEBI: CHEBI:82551 IUPAC Name: 1-ethenylpyrrolidin-2-one SMILES: *-CC(-*)N1CCCC1=O
Polyvinylpyrrolidone, M.W. 360,000
CAS: 9003-39-8 Molecular Formula: (C6H9NO)n Molecular Weight (g/mol): 111.14 MDL Number: MFCD01076626 InChI Key: WHNWPMSKXPGLAX-UHFFFAOYSA-N Synonym: n-vinyl-2-pyrrolidone,n-vinylpyrrolidone,1-vinyl-2-pyrrolidone,1-vinylpyrrolidin-2-one,n-vinyl-2-pyrrolidinone,vinylpyrrolidone,2-pyrrolidinone, 1-ethenyl,1-vinyl-2-pyrrolidinone,n-vinylpyrrolidinone,1-vinylpyrrolidone PubChem CID: 6917 ChEBI: CHEBI:82551 IUPAC Name: 1-ethenylpyrrolidin-2-one SMILES: *-CC(-*)N1CCCC1=O
Polyvinylpyrrolidone, average M.W. 3500, K12
CAS: 9003-39-8 Molecular Formula: (C6H9NO)n Molecular Weight (g/mol): 111.14 MDL Number: MFCD01076626 InChI Key: WHNWPMSKXPGLAX-UHFFFAOYSA-N Synonym: n-vinyl-2-pyrrolidone,n-vinylpyrrolidone,1-vinyl-2-pyrrolidone,1-vinylpyrrolidin-2-one,n-vinyl-2-pyrrolidinone,vinylpyrrolidone,2-pyrrolidinone, 1-ethenyl,1-vinyl-2-pyrrolidinone,n-vinylpyrrolidinone,1-vinylpyrrolidone PubChem CID: 6917 ChEBI: CHEBI:82551 IUPAC Name: 1-ethenylpyrrolidin-2-one SMILES: *-CC(-*)N1CCCC1=O
Polyvinylpyrrolidone, M.W. 8,000
CAS: 9003-39-8 Molecular Formula: (C6H9NO)n Molecular Weight (g/mol): 111.14 MDL Number: MFCD01076626 InChI Key: WHNWPMSKXPGLAX-UHFFFAOYSA-N Synonym: n-vinyl-2-pyrrolidone,n-vinylpyrrolidone,1-vinyl-2-pyrrolidone,1-vinylpyrrolidin-2-one,n-vinyl-2-pyrrolidinone,vinylpyrrolidone,2-pyrrolidinone, 1-ethenyl,1-vinyl-2-pyrrolidinone,n-vinylpyrrolidinone,1-vinylpyrrolidone PubChem CID: 6917 ChEBI: CHEBI:82551 IUPAC Name: 1-ethenylpyrrolidin-2-one SMILES: *-CC(-*)N1CCCC1=O
Polyvinylpyrrolidone K30, MP Biomedicals™
CAS: 9003-39-8 Molecular Formula: (C6H9NO)n Molecular Weight (g/mol): 111.14 MDL Number: MFCD01076626 InChI Key: WHNWPMSKXPGLAX-UHFFFAOYSA-N Synonym: n-vinyl-2-pyrrolidone,n-vinylpyrrolidone,1-vinyl-2-pyrrolidone,1-vinylpyrrolidin-2-one,n-vinyl-2-pyrrolidinone,vinylpyrrolidone,2-pyrrolidinone, 1-ethenyl,1-vinyl-2-pyrrolidinone,n-vinylpyrrolidinone,1-vinylpyrrolidone PubChem CID: 6917 ChEBI: CHEBI:82551 IUPAC Name: 1-ethenylpyrrolidin-2-one SMILES: *-CC(-*)N1CCCC1=O
Crospovidone (~40,000), TRC
CAS: 9003-39-8 Molecular Formula: (C6 H9 N O)mon Molecular Weight (g/mol): 111.14 Synonym: α-Hydro-ω-hydropoly[1-(2-oxopyrrolidin-1-yl)ethylene],2-Pyrrolidinone, 1-ethenyl-, homopolymer,2-Pyrrolidinone, 1-vinyl-, polymers (8CI),1-Vinyl-2-pyrrolidinone polymer,1-Vinyl-2-pyrrolidone homopolymer,1-Vinyl-2-pyrrolidone polymer,1-Vinylpyrrolidin-2-one homopolymer,143RP,40K,40K (vinyl polymer),7TS13,ACP 10,AT 717,Agent AT 717,Agrimer 15,Agrimer 30,Agrimer 90,Agrimer ATF,Agrimer K 30,Albigen A,Aldacol Q,Antaron P 804,Antitox Vana,Aqua 65JL,Aqua-Quench EC/WE,Arostit TL 0280,B 7509,Bolinan,Cevian A 88036,Creejus K 30,Creejus K 50,Creejus K 90,Crospovidone,Cross P.V.P.,Deltaspeed AF,Discol K 30L,Disintex 200,Divergan EF,Divergan F,Divergan RS,Epocros WS 560,Flexithix,Gaftex AE-K 15,Ganex P 804,HY 2,HY 3,Hemodesis,Hemodez,Hemodez N,Hemodez-N,Jarpol PVP-K 30,K 115,K 115 (vinyl polymer),K 12,K 120,K 15,K 15 (polymer),K 16-18,K 17,K 25,K 25 (surfactant),K 27/32,K 29-32,K 30,K 30C,K 30G,K 30PH,K 30W,K 40,K 50,K 50 (vinyl polymer),K 55,K 55 (vinyl polymer),K 60,K 60 (polymer),K 79,K 8,K 8 (vinyl polymer),K 85,K 85 (vinyl polymer),K 85N,K 85W,K 90,K 90HM,K 90W,K 92,K 92 (vinyl polymer),K88-96,Kollidon,Kollidon CE 50/50,Kollidon F 90,Kollidon K 12,Kollidon K 15,Kollidon K 17,Kollidon K 25,Kollidon K 29/32,Kollidon K 30,Kollidon K 60,Kollidon K 90,Kollidon K 90F,Kollidon R 90F,Konkrepol AD 1,Konkrepol B,LFC,Lumiten PPR 8450,Lumiten PR 8450,Luvicross M,Luviskol K,Luvitec K 15,Luvitec K 17,Luvitec K 30,Luvitec K 30PH,Luvitec K 40,Luvitec K 60,Luvitec K 80,Luvitec K 85,Luvitec K 90,Luvitec K 90PH,Luvitec PVP-K 30,MPK 90,MRTU 8457-79,N-Vinyl-2-pyrolidone homopolymer,N-Vinyl-2-pyrrolidinone homopolymer,N-Vinyl-2-pyrrolidone homopolymer,N-Vinyl-2-pyrrolidone polymer,N-Vinylbutyrolactam polymer,N-Vinylpyrrolidinone homopolymer,N-Vinylpyrrolidinone polymer,N-Vinylpyrrolidone homopolymer,N-Vinylpyrrolidone polymer,NKY-PVP-K,NP-K 30,NPK 15,NPK 90,Neocompensan,Neohemodes,P 0696,P-XL,PAK-K 15,PK 771,PV 03,PV 03 (vinyl pyrrolidone polymer),PV 05,PVK 30,PVP,PVP 10,PVP 1230,PVP 15,PVP 17PF,PVP 18,PVP 25,PVP 30,PVP 3000,PVP 360,PVP 360K,PVP 40,PVP 4000,PVP 40K,PVP 40T,PVP 50,PVP 99,PVP K 29/32,PVP-K,PVP-K 10,PVP-K 12,PVP-K 120,PVP-K 130,PVP-K 15,PVP-K 17,PVP-K 25,PVP-K 26/28,PVP-K 3,PVP-K 30,PVP-K 30W,PVP-K 35,PVP-K 40,PVP-K 45,PVP-K 60,PVP-K 64,PVP-K 70,PVP-K 78,PVP-K 79,PVP-K 80,PVP-K 85,PVP-K 88-96,PVP-K 90,PVP-K 90K95,PVP-K 95,PVPP,PVPP-XL 10,PX 4VP90,PX 90P,PX-K 30P,PX-K 90L,PX-K 90P,Peregal ST,Periston,Periston N,Peviston,Pitzcol K 120,Pitzcol K 17L,Pitzcol K 30,Pitzcol K 30L,Pitzcol K 50,Pitzcol K 90,Plasdone,Plasdone 4,Plasdone C,Plasdone XL,Plasdone XL 10,Plasmosan,Polidon,Poly(1-vinyl-2-pyrrolidinone),Poly(1-vinyl-2-pyrrolidone),Poly(1-vinylpyrrolidinone),Poly(N-vinyl-2-pyrrolidinone),Poly(N-vinyl-2-pyrrolidone),Poly(N-vinyl-2-pyrrolidone),Poly(N-vinyl-γ-butyrolactam),Poly(N-vinylbutyrolactam),Poly(N-vinylpyrrolidinone),Poly(N-vinylpyrrolidone),Poly(vinylpyrrolidinone),Poly(vinylpyrrolidone),Polyclar AT,Polyclar H,Polyclar L,Polyplasdone,Polyplasdone INF 10,Polyplasdone R-XL,Polyplasdone XL,Polyplasdone XL 10,Polyvidon,Polyvidon XL,Polyvidone,Polyvidone 25,Polyvidone K 30,Polyvinyl polypyrrolidone,Polyvinylpyrrolidon XL,Polyvinylpyrrolidone,Poviderm SK 3,Povidone,Povidone 30,Povidone K 12,Povidone K 15,Povidone K 17,Povidone K 25,Povidone K 29-32,Povidone K 30,Povidone K 90,Povidone K 90D,Protagent,Pvpon K 32,Rubiscole K 30,SD 13,SD 13 (polymer),Sokalan HP 165,Sokalan HP 50,Sokalan HP 53,Sokalan HP 60,Sokalan K 17P,Sokalan K 85P,Sokalan K 90,Sokalan K 90P,Sokolan HP 165,Sokolan HP 53,Sokolan HP 60,Sokolan K 30P,Sokolan K 85P,Subtosan,TSP 9A,Vinisil,Vinylpyrrolidinone polymer,Vinylpyrrolidone homopolymer,Vinylpyrrolidone polymer,ViviPrint 530,YK 001 IUPAC Name: 1-vinylpyrrolidin-2-one SMILES: C=CN1CCCC1=O
- 1
- 2