accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 9003-53-6

IUPAC Name: ethenylbenzene
Synonyms: styrene fema number 3234 bulstren K-525-19 styron styrol styreen styrole ccris 564 styropol SO styrolene
Molecular Weight (g/mol): 104.15
Molecular Formula: C8H8
InChi Key: PPBRXRYQALVLMV-UHFFFAOYSA-N
SMILES: C=CC1=CC=CC=C1
Quantity 
  • (14)
  • (1)
  • (2)
  • (1)
  • (2)
  • (9)
  • (1)
  • (1)
  • (2)
  • (9)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (40)
  • (3)
  • (2)
  • (14)
  • (1)
  • (2)
  • (1)
  • (2)
  • (9)
  • (1)
  • (1)
  • (2)
  • (9)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)