accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

A(a)

CAS RN 9003-69-4

IUPAC Name: 1,2-diethenylbenzene
Synonyms: divinylbenzene
Molecular Weight (g/mol): 130.19
Molecular Formula: C10H10
InChi Key: MYRTYDVEIRVNKP-UHFFFAOYSA-N
SMILES: C=CC1=CC=CC=C1C=C
Color 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)