accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

A(a)

CAS RN 9003-70-7

IUPAC Name: tert-butyl 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoate
Synonyms: tert-butyl 4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-oxopentanoate
Molecular Weight (g/mol): 409.48
Molecular Formula: C24H27NO5
InChi Key: BCIPGSZQUDLGSY-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C=O
Quantity 
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (3)
Form 
  • (3)
Product Type 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (3)
  • (2)
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)