accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

O∗O∗OORORORORORORn

CAS RN 9004-32-4

IUPAC Name: 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
Synonyms: cellulose gum
Molecular Weight (g/mol): 263.2
Molecular Formula: C8H16NaO8
InChi Key: DPXJVFZANSGRMM-UHFFFAOYNA-N
SMILES: [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
Molecular Weight (g/mol) 
  • (3)
  • (20)
Quantity 
  • (7)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
Percent Purity 
  • (11)
  • (1)
Physical Form 
  • (6)
  • (1)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (11)
  • (2)
  • (3)
  • (5)
  • (3)
  • (20)
  • (7)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (11)
  • (1)
  • (6)
  • (1)
  • (5)