accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 9004-34-6

IUPAC Name: 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol
Synonyms: 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol
Molecular Weight (g/mol): 370.35
Molecular Formula: C14H26O11
InChi Key: PTHCMJGKKRQCBF-UHFFFAOYNA-N
SMILES: COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (6)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)