accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 9004-35-7

O(R)(S)O(R)(R)(S)OOOHH3COnR1

CAS RN 9004-35-7

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula: (C6H7O5)nC2H3O
InChi Key:
SMILES:
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)