CAS RN 9004-67-5
Thermo Scientific Chemicals Methyl cellulose, viscosity 400 cPs
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl cellulose, viscosity 15 cPs
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Thermo Scientific Chemicals Methyl cellulose, viscosity 4000 cPs
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Thermo Scientific Chemicals Methyl cellulose, viscosity 4000 cP (2% solution in water)
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl cellulose, viscosity 400 cP (2% solution in water)
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Thermo Scientific Chemicals Methyl cellulose, viscosity 15 cP (2% solution in water)
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl cellulose, viscosity 1600 cPs
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Thermo Scientific Chemicals Methyl cellulose, viscosity 8000 cPs
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 MDL Number: MFCD00081763 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl cellulose, MP Biomedicals™
CAS: 9004-67-5 Molecular Formula: C20H38O11 MDL Number: MFCD00081763 Synonym: Cellulose methyl ether,Methocel MC,Cellothyl,Syncelose PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl Cellulose, viscosity of 2% aqueous solution (25deg.C) 1,500cps, MP Biomedicals
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N Synonym: Cellulose methyl ether PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methyl Cellulose, viscosity of 2% aqueous solution (25°C) 15cps, MP Biomedicals
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.513 InChI Key: YLGXILFCIXHCMC-JHGZEJCSSA-N Synonym: Cellulose methyl ether PubChem CID: 51063134 IUPAC Name: (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC
Methylcellulose, TRC
CAS: 9004-67-5 Molecular Formula: C20H38O11 Molecular Weight (g/mol): 454.5 Synonym: Adulsin,Avicel SG,Bagolax,Benecel 143HR,Benecel M 0,Benecel M 011,Benecel M 02,Benecel M 021,Benecel M 043,Benecel MC 4000PS,Benecel MO 42,Bufapto Methalose,Bulkaloid,Celacol M;,Methocel 4C,Methocel 64625,Methocel A,Methocel A 100,Methocel A 125LV,Methocel A 15,Methocel A 15C,Methocel A 15C Premium,Methocel A 15FG,Methocel A 15LV,Methocel A 15LV Premium,Methocel A 40M,Methocel A 4C,Methocel A 4C Premium,Methocel A 4C-FG,Methocel A 4M,Methocel A 4M Premium,Methocel A 50,Methocel AC Premium,Methocel CHG,Methocel Fluka 64625,Methocel HPM 500DS,Methocel HPM 50DS,Metolose SM 100,Metolose SM 10000,Metolose SM 15,Metolose SM 1500,Metolose SM 25,Metolose SM 4,Metolose SM 400,Metolose SM 4000,Metolose SM 65-15,Metolose SM 800,Metolose SM 8000,Metolose SR,Napolone,Nicel,O-Methylcellulose,Probe D,Rhomellose,SBQ 30000PE,SM 100,SM 10000,SM 100000,SM 15Tylose SAP,Tylose SL,Tylose SL 100,Tylose SL 400,Tylose SL 600,Tylose TWA,V 32,V 32 (cellulose derivative),Viscol,Viscontran L 52,Viscontran MC 400,Viscosol,Viskontran MC 10S,Walocel M,Walocel M 20678,Walsroder MC 20000S,XT 6000 SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC