accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 9004-98-2

IUPAC Name: 2-[(9Z)-octadec-9-en-1-yloxy]ethan-1-ol
Synonyms: oleth-2 oleth-5 oleth-6 oleth-7 oleth-8 oleth-3 oleth-4 oleth-9 oleth-106 oleth-16
Molecular Weight (g/mol): 312.54
Molecular Formula: C20H40O2
InChi Key: KWVPFECTOKLOBL-KTKRTIGZSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCOCCO
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results