accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 9005-38-3

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula: (C6H7O7)A(C6H7O7)BNa
InChi Key:
SMILES: [Na+].O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C([O-])=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C([O-])=O
Quantity 
  • (3)
  • (7)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (6)
Physical Form 
  • (11)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (3)
  • (5)
  • (3)
  • (7)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (6)
  • (11)
  • (1)