accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 9005-65-6

IUPAC Name: 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
Synonyms: polysorbate 81 peg-3 sorbitan oleate peg-20 sorbitan oleate peg-40 sorbitan oleate peg-6 sorbitan oleate
Molecular Weight (g/mol): 604.82
Molecular Formula: C32H60O10
InChi Key: RGPBUVUVZKQNHD-MDZDMXLPNA-N
SMILES: CCCCCCCC\C=C\CCCCCCCC(=O)OCCOCC(OCCO)C1OC(CC1OCCO)OCCO
Molecular Weight (g/mol) 
  • (3)
  • (18)
Quantity 
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
Grade 
  • (8)
pH 
  • (1)
  • (3)
  • (1)
  • (2)
Form 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (8)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (3)
  • (7)
  • (3)
  • (18)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (8)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)