accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 9005-66-7

IUPAC Name: 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
Synonyms: peg-80 sorbitan laurate sorbimacrogol laurate 300 polysorbate 21 polysorbate 61 polyethylene glycol (80) sorbitan monolaurate polyoxyethylene (80) sorbitan monolaurate crillet 2 polyoxyethylene (4) sorbitan monostearate polyoxyethylene (44) sorbitan monolaurate protasorb L-20
Molecular Weight (g/mol): 522.68
Molecular Formula: C26H50O10
InChi Key: HMFKFHLTUCJZJO-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
pH 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results