CAS RN 90098-06-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Deschloro-2-chlorobenzoyl Rebapimide, TRC
CAS: 90098-06-9 Molecular Formula: C19H15ClN2O4 Molecular Weight (g/mol): 370.79 Synonym: alpha-[(2-Chlorobenzoyl)amino]-1,2-dihydro-2-oxo-4-Quinolinepropanoic Acid; IUPAC Name: 2-[(2-chlorobenzoyl)amino]-3-(2-oxo-1H-quinolin-4-yl)propanoic acid SMILES: OC(=O)C(CC1=CC(=O)Nc2ccccc12)NC(=O)c3ccccc3Cl