accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 90273-30-6

IUPAC Name: 5-methyl-1-benzothiophene-2-sulfonyl chloride
Synonyms: 5-methyl-1-benzothiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 246.72
Molecular Formula: C9H7ClO2S2
InChi Key: YGGJYXHTCDQVDB-UHFFFAOYSA-N
SMILES: CC1=CC=C2SC(=CC2=C1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results