accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 902836-42-4

HNNNCl

CAS RN 902836-42-4

IUPAC Name: 4-[4-(2-chlorophenyl)-1H-pyrazol-1-yl]piperidine
Synonyms: 4-[4-(2-chlorophenyl)pyrazol-1-yl]piperidine
Molecular Weight (g/mol): 261.75
Molecular Formula: C14H16ClN3
InChi Key: RRRIXLCXVVGUTG-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C1=CN(N=C1)C1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results