accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NONOHO

CAS RN 902837-22-3

IUPAC Name: 3-(3-ethyl-1,2,4-oxadiazol-5-yl)benzoic acid
Synonyms: 3-(3-ethyl-1,2,4-oxadiazol-5-yl)benzoic acid
Molecular Weight (g/mol): 218.21
Molecular Formula: C11H10N2O3
InChi Key: UDGPERFBSLKMSD-UHFFFAOYSA-N
SMILES: CCC1=NOC(=N1)C1=CC(=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results