accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 902837-44-9

H3CH3CCH3OONHNN

CAS RN 902837-44-9

IUPAC Name: tert-butyl N-(5-cyanopyridin-2-yl)carbamate
Synonyms: tert-butyl N-(5-cyanopyridin-2-yl)carbamate
Molecular Weight (g/mol): 219.24
Molecular Formula: C11H13N3O2
InChi Key: JDQQNZYKHZVLAM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=NC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results