accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOHNH2SSOOHNH2HClHCl

CAS RN 90350-38-2

IUPAC Name: 2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid dihydrochloride
Synonyms: cystine dihydrochloride
Molecular Weight (g/mol): 313.21
Molecular Formula: C6H14Cl2N2O4S2
InChi Key: HHGZUQPEIHGQST-UHFFFAOYNA-N
SMILES: Cl.Cl.NC(CSSCC(N)C(O)=O)C(O)=O
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results