accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH2ONHNFFF

CAS RN 90357-53-2

IUPAC Name: N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide
Synonyms: N-[4-cyano-3-(trifluoromethyl)phenyl]-2-methylprop-2-enamide
Molecular Weight (g/mol): 254.21
Molecular Formula: C12H9F3N2O
InChi Key: HHWDZLSGDDXUSM-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)NC1=CC(=C(C=C1)C#N)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)