accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 90390-27-5

IUPAC Name: 1-(3,5-difluorophenyl)methanamine
Synonyms: 1-(3,5-difluorophenyl)methanamine
Molecular Weight (g/mol): 143.14
Molecular Formula: C7H7F2N
InChi Key: VJNGGOMRUHYAMC-UHFFFAOYSA-N
SMILES: NCC1=CC(F)=CC(F)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results