accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 90396-06-8

IUPAC Name: 6-chloro-3,4-dihydro-2H-1λ⁶-benzothiopyran-1,1,4-trione
Synonyms: 6-chloro-2,3-dihydro-1λ⁶-benzothiopyran-1,1,4-trione
Molecular Weight (g/mol): 230.66
Molecular Formula: C9H7ClO3S
InChi Key: ZFZPMYLZPMMUQA-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C(=C1)C(=O)CCS2(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results