accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 90536-66-6

IUPAC Name: 2-(4-methanesulfonylphenyl)acetate
Synonyms: 2-(4-methanesulfonylphenyl)acetate
Molecular Weight (g/mol): 213.23
Molecular Formula: C9H9O4S
InChi Key: HGGWOSYNRVOQJH-UHFFFAOYSA-M
SMILES: CS(=O)(=O)C1=CC=C(CC([O-])=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results