CAS RN 906352-89-4
IUPAC Name:
5-(2-bromophenyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole
Synonyms:
5-(2-bromophenyl)-1-methyl-3-(trifluoromethyl)pyrazole
Molecular Weight (g/mol):
305.1
Molecular Formula:
C11H8BrF3N2
InChi Key:
UKDUPCWWWMHDRH-UHFFFAOYSA-N
SMILES:
CN1N=C(C=C1C1=CC=CC=C1Br)C(F)(F)F
1
–
1
of
1
results
1
5-(2-Bromophenyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole, ≥97%, Thermo Scientific™
CAS: 906352-89-4 Molecular Formula: C11H8BrF3N2 Molecular Weight (g/mol): 305.098 MDL Number: MFCD09817512 InChI Key: UKDUPCWWWMHDRH-UHFFFAOYSA-N Synonym: 5-2-bromophenyl-1-methyl-3-trifluoromethyl-1h-pyrazole,5-2-bromophenyl-1-methyl-3-trifluoromethyl pyrazole PubChem CID: 24229660 IUPAC Name: 5-(2-bromophenyl)-1-methyl-3-(trifluoromethyl)pyrazole SMILES: CN1C(=CC(=N1)C(F)(F)F)C2=CC=CC=C2Br