accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 906353-04-6

IUPAC Name: 2-(morpholin-4-yl)-1,3-thiazole-4-carboxylic acid
Synonyms: 2-(morpholin-4-yl)-1,3-thiazole-4-carboxylic acid
Molecular Weight (g/mol): 214.24
Molecular Formula: C8H10N2O3S
InChi Key: JXQGQYNBVNKCAK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CSC(=N1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results