CAS RN 908266-48-8
CAS RN 908266-48-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tropine-3-thiol Hydrochloride, TRC
CAS: 908266-48-8 Chemical Name or Material: Tropine-3-thiol Hydrochloride Formula Weight: 193.0692 InChI Formula: InChI=1S/C8H15NS.ClH/c1-9-6-2-3-7(9)5-8(10)4-6;/h6-8,10H,2-5H2,1H3;1H/t6-,7?,8+;/m0./s1 IUPAC Name: (5S)-8-methyl-8-azabicyclo[3.2.1]octane-3-thiol;hydrochloride Molecular Formula: C8H16ClNS Molecular Weight (g/mol): 193.74 Recommended Storage: 4°C SMILES: Cl.CN1[C@H]2CCC1C[C@H](S)C2 Synonym: (3-endo)-8-Methyl-8-Azabicyclo[3.2.1]octane-3-thiol Hydrochloride