accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 90843-31-5

IUPAC Name: 1-(2,3-dihydro-1-benzofuran-5-yl)ethan-1-one
Synonyms: 1-(2,3-dihydro-1-benzofuran-5-yl)ethanone
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: MMVUJVASBDVNGJ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2OCCC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results