accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFMgBr

CAS RN 90897-92-0

IUPAC Name: magnesium(2+) 3,4-difluorobenzen-1-ide bromide
Synonyms: magnesium(2+) 3,4-difluorobenzen-1-ide bromide
Molecular Weight (g/mol): 217.3
Molecular Formula: C6H3BrF2Mg
InChi Key: AULUGBNIKQAHPF-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].FC1=CC=[C-]C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results