accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNNNNSNH

CAS RN 909910-43-6

IUPAC Name: 3-(6-methylpyridin-2-yl)-N-phenyl-4-(quinolin-4-yl)-1H-pyrazole-1-carbothioamide
Synonyms: 3-(6-methylpyridin-2-yl)-N-phenyl-4-(quinolin-4-yl)pyrazole-1-carbothioamide
Molecular Weight (g/mol): 421.52
Molecular Formula: C25H19N5S
InChi Key: HIJMSZGHKQPPJS-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=N1)C1=NN(C=C1C1=C2C=CC=CC2=NC=C1)C(=S)NC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results