accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 91-08-7

IUPAC Name: 1,3-diisocyanato-2-methylbenzene
Synonyms: 2,6-toluene diisocyanate m-tolylene diisocyanate meta-tolylene diisocyanate
Molecular Weight (g/mol): 174.16
Molecular Formula: C9H6N2O2
InChi Key: RUELTTOHQODFPA-UHFFFAOYSA-N
SMILES: CC1=C(C=CC=C1N=C=O)N=C=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)