accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3

CAS RN 91-16-7

IUPAC Name: 1,2-dimethoxybenzene
Synonyms: veratrole methyl guaiacol 2-dimethoxybenzol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: ABDKAPXRBAPSQN-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1OC
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (3)
  • (3)
  • (3)