accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 91-21-4

IUPAC Name: 1,2,3,4-tetrahydroisoquinoline
Synonyms: tetrahydroisoquinoline
Molecular Weight (g/mol): 133.19
Molecular Formula: C9H11N
InChi Key: UWYZHKAOTLEWKK-UHFFFAOYSA-N
SMILES: C1CC2=CC=CC=C2CN1
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (5)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results