accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

N

CAS RN 91-22-5

IUPAC Name: quinoline
Synonyms: cinch benzopyridine benzo(b)pyridine chinoleine usaf EK-218 leukol leucol 2,3-benzopyridine ccris 547 quinoline (substance)
Molecular Weight (g/mol): 129.16
Molecular Formula: C9H7N
InChi Key: SMWDFEZZVXVKRB-UHFFFAOYSA-N
SMILES: C1=CC=C2N=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (6)
  • (1)
  • (7)
Percent Purity 
  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
Physical Form 
  • (14)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (8)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (7)
  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (14)
  • (1)
  • (1)
  • (1)