accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 91-64-5

IUPAC Name: 2H-chromen-2-one
Synonyms: 2H-1-benzopyran-2-one coumarinic lactone 2-benzopyrone 1 o-hydroxyzimtsaure-lacton venalot mono epa pesticide chemical code 127301 rattex α benzopyrone o-hydroxycinnamic acid lactone coumarinic anhydride
Molecular Weight (g/mol): 146.15
Molecular Formula: C9H6O2
InChi Key: ZYGHJZDHTFUPRJ-UHFFFAOYSA-N
SMILES: O=C1OC2=CC=CC=C2C=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)