accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 91000-53-2

HNOHOHOHSOOONa

CAS RN 91000-53-2

IUPAC Name: sodium 3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propane-1-sulfonate
Synonyms: sodium 3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propane-1-sulfonate
Molecular Weight (g/mol): 265.26
Molecular Formula: C7H16NNaO6S
InChi Key: FEGYIWVHCSRXCG-UHFFFAOYSA-M
SMILES: [Na+].OCC(CO)(CO)NCCCS([O-])(=O)=O
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Concentration 
  • (2)
  • (1)
pH 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results