accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 910036-92-9

IUPAC Name: 1-[3-(pyrimidin-2-yl)phenyl]methanamine
Synonyms: 1-[3-(pyrimidin-2-yl)phenyl]methanamine
Molecular Weight (g/mol): 185.23
Molecular Formula: C11H11N3
InChi Key: MXSXDKVANPZCBQ-UHFFFAOYSA-N
SMILES: NCC1=CC=CC(=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results