accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HNONCH3CH3

CAS RN 910037-06-8

IUPAC Name: dimethyl(3-{2-[(methylamino)methyl]phenoxy}propyl)amine
Synonyms: dimethyl(3-{2-[(methylamino)methyl]phenoxy}propyl)amine
Molecular Weight (g/mol): 222.33
Molecular Formula: C13H22N2O
InChi Key: PSLNWMDOKBMVNV-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC=C1OCCCN(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results