accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 910037-15-9

IUPAC Name: 4-methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H,3H,4H-pyrido[3,2-b][1,4]oxazine
Synonyms: 4-methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H,3H-pyrido[3,2-b][1,4]oxazine
Molecular Weight (g/mol): 276.14
Molecular Formula: C14H21BN2O3
InChi Key: PDTRXHRSCGNYFD-UHFFFAOYSA-N
SMILES: CN1CCOC2=C1N=CC(=C2)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results