accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 91062-38-3

BOOCH3OHHO

CAS RN 91062-38-3

IUPAC Name: [4-(propoxycarbonyl)phenyl]boronic acid
Synonyms: 4-(propoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 208.02
Molecular Formula: C10H13BO4
InChi Key: XJEONFAJOXSZDX-UHFFFAOYSA-N
SMILES: CCCOC(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results