accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 91076-93-6

IUPAC Name: methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate
Synonyms: methyl 3-amino-5-(4-chlorophenyl)thiophene-2-carboxylate
Molecular Weight (g/mol): 267.73
Molecular Formula: C12H10ClNO2S
InChi Key: MELGGDOYSMRBGA-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C=C(S1)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results