accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNNHFFFHCl

CAS RN 911064-58-9

IUPAC Name: 2-(trifluoromethyl)-5H,6H,7H,8H-imidazo[1,2-a]pyrazine hydrochloride
Synonyms: 2-(trifluoromethyl)-5H,6H,7H,8H-imidazo[1,2-a]pyrazine hydrochloride
Molecular Weight (g/mol): 227.62
Molecular Formula: C7H9ClF3N3
InChi Key: WQJNJYFZTOYWPU-UHFFFAOYSA-N
SMILES: Cl.FC(F)(F)C1=CN2CCNCC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results